Anti-METAP1 Mouse Monoclonal Antibody
Catalog # ABGEAM1922B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Methionyl aminopeptidase 1
- Antigen symbol:METAP1
- Clonality:Monoclonal
- Clone:248CT14.6.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Gene ID:23173
- Antigen synonyms:MetAP1A|MAP1A
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:43.215 kDa
- Sequence:SESEQALKGTSQIKLLSSEDIEGMRLVCRLAREVLDVAAGMIKPGVTTEEIDHAVHLACIARNCYPSPLNYYNFPKSCCTSVNEVICHGIPDRRPLQEGDIVNVDITLYRNGYHGDLNETFFVGEVDDGARKLVQTTYECLMQAIDAVKPGVRYRELGNIIQKHAQANGFSVVRSYCGHGIHKLFHTAPNVPHYAKNKAVGVMKSGHVFTIEPMICEGGWQDETWPDGWTAVTRDGKRSAQFEHTLLVTDTGCEILTRRLDSARPHFMSQF
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This METAP1 monoclonal antibody is generated from mouse immunized with METAP1 recombinant protein.
- Purification:Protein G Purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: METAP1 (methionyl aminopeptidase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human