Anti-OR5V1 Rabbit Polyclonal Antibody
AVIVARP68977_P050
- Antibody type:Primary
- Antigen name:olfactory receptor, family 5, subfamily V, member 1
- Antigen symbol:OR5V1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:81696
- Antigen synonyms:hs6M1-21|6M1-21
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: OR5V1 (olfactory receptor, family 5, subfamily V, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human