Order Entry
Italy
ContactUsLinkComponent
Anti-PPP1R12C Rabbit Polyclonal Antibody
Anti-PPP1R12C Rabbit Polyclonal Antibody
Catalog # AVIVARP67757_P050
undefined
Anti-PPP1R12C Rabbit Polyclonal Antibody
Catalog # AVIVARP67757_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    protein phosphatase 1, regulatory subunit 12C
  • Antigen symbol:
    PPP1R12C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    54776
  • Antigen synonyms:
    p84|p85|LENG3|MBS85
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: PPP1R12C (protein phosphatase 1, regulatory subunit 12C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human