Anti-FGFR1OP2 Rabbit Polyclonal Antibody
AVIVARP65755_P050
- Antibody type:Primary
- Antigen name:FGFR1 oncogene partner 2
- Antigen symbol:FGFR1OP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:26127
- Antigen synonyms:HSPC123-like|WIT3.0
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: FGFR1OP2 (FGFR1 oncogene partner 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human