Order Entry
Italy
ContactUsLinkComponent
Anti-PDE7A Rabbit Polyclonal Antibody
Anti-PDE7A Rabbit Polyclonal Antibody
  AVIVARP64421_P050
undefined
Anti-PDE7A Rabbit Polyclonal Antibody
  AVIVARP64421_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    phosphodiesterase 7A
  • Antigen symbol:
    PDE7A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    5150
  • Antigen synonyms:
    PDE7|HCP1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: PDE7A (phosphodiesterase 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human