Anti-PRKCSH Rabbit Polyclonal Antibody
Catalog # AVIVARP64007_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase C Substrate 80K-H
- Antigen symbol:PRKCSH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5589
- Antigen synonyms:AGE-R2|PLD1|PKCSH|G19P1|PCLD
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PRKCSH (protein kinase C substrate 80K-H)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human