Anti-RPS3 Rabbit Polyclonal Antibody
AVIVARP63709_P050
- Antibody type:Primary
- Antigen name:Ribosomal Protein S3
- Antigen symbol:RPS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:6188
- Antigen synonyms:S3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RPS3 (ribosomal protein S3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: