Anti-NPL Rabbit Polyclonal Antibody
Catalog # AVIVARP63210_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase)
- Antigen symbol:NPL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:80896
- Antigen synonyms:NPL1|NAL|C1orf13|C112
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NPL (N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: