Anti-BMP4 Rabbit Polyclonal Antibody
Catalog # AVIVARP63062_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:bone morphogenetic protein 4
- Antigen symbol:BMP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:652
- Antigen synonyms:BMP2B1|MCOPS6|ZYME|OFC11|BMP2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: BMP4 (bone morphogenetic protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: