Anti-NQO1 Rabbit Polyclonal Antibody
Catalog # AVIVARP61165_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:NAD(P)H dehydrogenase, quinone 1
- Antigen symbol:NQO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1728
- Antigen synonyms:NMORI|NMOR1|DIA4|DTD|DHQU|QR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: