Anti-OR2H2 Rabbit Polyclonal Antibody
Catalog # AVIVARP59730_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:olfactory receptor, family 2, subfamily H, member 2
- Antigen symbol:OR2H2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:7932
- Antigen synonyms:OR2H3|dJ271M21.2|OLFR42B|OLFR2|hs6M1-12|FAT11
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OR2H2 (olfactory receptor, family 2, subfamily H, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human