Anti-GNL3L Rabbit Polyclonal Antibody
AVIVARP58798_P050
- Antibody type:Primary
- Antigen name:Guanine nucleotide binding protein-like 3 (nucleolar)-like
- Antigen symbol:GNL3L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:54552
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GNL3L (guanine nucleotide binding protein-like 3 (nucleolar)-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat