Anti-EPHA5 Rabbit Polyclonal Antibody
Catalog # AVIVARP58460_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:EPH receptor A5
- Antigen symbol:EPHA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:2044
- Antigen synonyms:TYRO4|EHK-1|CEK7|EHK1|HEK7|EK7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EPHA5 (EPH receptor A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: