Anti-TFDP3 Rabbit Polyclonal Antibody
Catalog # AVIVARP58170_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:transcription factor Dp family, member 3
- Antigen symbol:TFDP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:51270
- Antigen synonyms:DP4|HCA661|CT30
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TFDP3 (transcription factor Dp family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human