Anti-OSER1 Rabbit Polyclonal Antibody
Catalog # AVIVARP56915_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:oxidative stress responsive serine-rich 1
- Antigen symbol:OSER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:51526
- Antigen synonyms:HSPC207|Perit1|Osr1|dJ1183I21.1|C20orf111
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OSER1 (oxidative stress responsive serine-rich 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: