Order Entry
Italy
ContactUsLinkComponent
Anti-FGF21 Rabbit Polyclonal Antibody
Anti-FGF21 Rabbit Polyclonal Antibody
Catalog # AVIVARP55368_P050
undefined
Anti-FGF21 Rabbit Polyclonal Antibody
Catalog # AVIVARP55368_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor 21
  • Antigen symbol:
    FGF21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    26291
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: FGF21 (fibroblast growth factor 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat