Anti-THYN1 Rabbit Polyclonal Antibody
Catalog # AVIVARP54987_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Thymocyte Nuclear Protein 1
- Antigen symbol:THYN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Guinea pig,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:29087
- Antigen synonyms:THY28KD|MDS012|HSPC144|MY105|THY28
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: THYN1 (thymocyte nuclear protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Guinea pig, Human, Mouse, Rat