Anti-RP11-269F19.9 Rabbit Polyclonal Antibody
AVIVARP54425_P050
- Antibody type:Primary
- Antigen name:Tctex1 domain containing 4
- Antigen symbol:TCTEX1D4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:343521
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RP11-269F19.9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: