Anti-METTL21A Rabbit Polyclonal Antibody
AVIVARP52844_P050
- Antibody type:Primary
- Antigen name:Methyltransferase like 21A
- Antigen symbol:METTL21A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:151194
- Antigen synonyms:FAM119A|HSPA-KMT|HCA557b
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: METTL21A (methyltransferase like 21A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat