Order Entry
Italy
ContactUsLinkComponent
Anti-ATP5H Rabbit Polyclonal Antibody
Anti-ATP5H Rabbit Polyclonal Antibody
  AVIVARP52160_P050
Anti-ATP5H Rabbit Polyclonal Antibody
  AVIVARP52160_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d
  • Antigen symbol:
    ATP5H
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    10476
  • Antigen synonyms:
    ATPQ
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: ATP5H (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: