Order Entry
Italy
ContactUsLinkComponent
Anti-PYROXD2 Rabbit Polyclonal Antibody
Anti-PYROXD2 Rabbit Polyclonal Antibody
  AVIVARP51276_P050
Anti-PYROXD2 Rabbit Polyclonal Antibody
  AVIVARP51276_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    pyridine nucleotide-disulphide oxidoreductase domain 2
  • Antigen symbol:
    PYROXD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    84795
  • Antigen synonyms:
    FP3420|C10orf33
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: PYROXD2 (pyridine nucleotide-disulphide oxidoreductase domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human