Anti-SPOP Rabbit Polyclonal Antibody
Catalog # AVIVARP50348_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:speckle-type POZ protein
- Antigen symbol:SPOP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:8405
- Antigen synonyms:BTBD32|TEF2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SPOP (speckle-type POZ protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: