Order Entry
Italy
ContactUsLinkComponent
 
Anti-TREML2 Rabbit Polyclonal Antibody
  AVIVARP49877_P050
Anti-TREML2 Rabbit Polyclonal Antibody
  AVIVARP49877_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Triggering receptor expressed on myeloid cells-like 2
  • Antigen symbol:
    TREML2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    79865
  • Antigen synonyms:
    C6orf76|dJ238O23.1|TLT2|TLT-2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: TREML2 (triggering receptor expressed on myeloid cells-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: