Order Entry
Italy
ContactUsLinkComponent
Anti-SCUBE2 Rabbit Polyclonal Antibody
Anti-SCUBE2 Rabbit Polyclonal Antibody
Catalog # AVIVARP49604_P050
Anti-SCUBE2 Rabbit Polyclonal Antibody
Catalog # AVIVARP49604_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Signal peptide, CUB domain, EGF-like 2
  • Antigen symbol:
    SCUBE2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    57758
  • Antigen synonyms:
    CEGP1|CEGB1|CEGF1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: SCUBE2 (signal peptide, CUB domain, EGF-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: