Anti-SEMA6A Rabbit Polyclonal Antibody
Catalog # AVIVARP49574_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 6A
- Antigen symbol:SEMA6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:57556
- Antigen synonyms:HT018|SEMAQ|SEMA|VIA|SEMA6A1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SEMA6A (sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: