Anti-GALNT16 Rabbit Polyclonal Antibody
Catalog # AVIVARP49540_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Polypeptide N-Acetylgalactosaminyltransferase 16
- Antigen symbol:GALNT16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:57452
- Antigen synonyms:GALNACT16|GALNTL1|GalNAc-T16
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GALNT16 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: