Anti-KIAA0101 Rabbit Polyclonal Antibody
Catalog # AVIVARP46033_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen symbol:KIAA0101
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:9768
- Antigen synonyms:PAF15|PAF|L5|p15(PAF)|OEATC|p15PAF|NS5ATP9|p15/PAF|OEATC1|OEATC-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KIAA0101 (KIAA0101)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: