Anti-DONSON Rabbit Polyclonal Antibody
AVIVARP45862_P050
- Antibody type:Primary
- Antigen name:downstream neighbor of SON
- Antigen symbol:DONSON
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:29980
- Antigen synonyms:C21orf60|B17
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DONSON (downstream neighbor of SON)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: