Anti-ITGA7 Rabbit Polyclonal Antibody
AVIVARP45241_P050
- Antibody type:Primary
- Antigen name:Integrin Alpha 7
- Antigen symbol:ITGA7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:81008
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ITGA7 (integrin, alpha 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: