Anti-WLS Rabbit Polyclonal Antibody
AVIVARP44430_P050
- Antibody type:Primary
- Antigen name:wntless Wnt ligand secretion mediator
- Antigen symbol:WLS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:79971
- Antigen synonyms:mig-14|EVI|C1orf139|GPR177|MRP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: WLS (wntless Wnt ligand secretion mediator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat