Order Entry
Italy
ContactUsLinkComponent
Anti-FBXO24 Rabbit Polyclonal Antibody
Anti-FBXO24 Rabbit Polyclonal Antibody
Catalog # AVIVARP43114_P050
Anti-FBXO24 Rabbit Polyclonal Antibody
Catalog # AVIVARP43114_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    F-box protein 24
  • Antigen symbol:
    FBXO24
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    26261
  • Antigen synonyms:
    FBX24
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: FBXO24 (F-box protein 24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human