Anti-FBXW2 Rabbit Polyclonal Antibody
Catalog # AVIVARP43109_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:F-box and WD repeat domain containing 2
- Antigen symbol:FBXW2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:26190
- Antigen synonyms:FBW2|Md6|Fwd2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FBXW2 (F-box and WD repeat domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: