Anti-RAPGEF2 Rabbit Polyclonal Antibody
Catalog # AVIVARP42759_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Rap guanine nucleotide exchange factor (GEF) 2
- Antigen symbol:RAPGEF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RAPGEF2 (Rap guanine nucleotide exchange factor (GEF) 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: