Anti-FNBP1 Rabbit Polyclonal Antibody
AVIVARP40800_P050
- Antibody type:Primary
- Antigen name:Formin Binding Protein 1
- Antigen symbol:FNBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:14269
- Antigen synonyms:1110057E06Rik|Fbp17|FBP1|2210010H06Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: FNBP1 (formin binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: