Anti-PHF21A Rabbit Polyclonal Antibody
Catalog # AVIVARP39269_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:PHD finger protein 21A
- Antigen symbol:PHF21A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:51317
- Antigen synonyms:BHC80|BM-006
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PHF21A (PHD finger protein 21A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: