Anti-MEF2D Rabbit Polyclonal Antibody
AVIVARP38699_P050
- Antibody type:Primary
- Antigen name:Myocyte enhancer factor 2D
- Antigen symbol:MEF2D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:4209
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MEF2D (myocyte enhancer factor 2D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: