Anti-TAF6 Rabbit Polyclonal Antibody
Catalog # AVIVARP38652_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa
- Antigen symbol:TAF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6878
- Antigen synonyms:MGC:8964|TAF(II)80|TAFII70|TAFII85|TAFII-80|TAFII80|TAF(II)70|TAFII-70|TAF2E
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TAF6 (TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: