Anti-FOXG1 Rabbit Polyclonal Antibody
Catalog # AVIVARP38596_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box G1
- Antigen symbol:FOXG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:2290
- Antigen synonyms:KHL2|HBF-3|QIN|HBF-G2|FOXG1A|HFK1|HFK2|HFK3|FKHL4|FKHL3|FKH2|BF2|FHKL3|FKHL1|HBF2|HBF-2|FOXG1B|BF1|FOXG1C|HBF-1|FKHL2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FOXG1 (forkhead box G1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat