Order Entry
Italy
ContactUsLinkComponent
Anti-ELK3 Rabbit Polyclonal Antibody
Anti-ELK3 Rabbit Polyclonal Antibody
  AVIVARP38590_P050
undefined
Anti-ELK3 Rabbit Polyclonal Antibody
  AVIVARP38590_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    ELK3, member of ETS oncogene family
  • Antigen symbol:
    ELK3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    13713
  • Antigen synonyms:
    D430049E23Rik|Net|Erp|Etrp|Sap-2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: ELK3 (ELK3, ETS-domain protein (SRF accessory protein 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: