Anti-ASCL2 Rabbit Polyclonal Antibody
AVIVARP36912_T100
- Antibody type:Primary
- Antigen name:Achaete-scute Complex Homolog 2
- Antigen symbol:ASCL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:17173
- Antigen synonyms:2410083I15Rik|bHLHa45|Mash2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: ASCL2 (achaete-scute family bHLH transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: