Anti-ZNF490 Rabbit Polyclonal Antibody
AVIVARP35978_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 490
- Antigen symbol:ZNF490
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:57474
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF490 (zinc finger protein 490)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human