Anti-LHX9 Rabbit Polyclonal Antibody
Catalog # AVIVARP35965_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:LIM homeobox 9
- Antigen symbol:LHX9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:56956
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: LHX9 (LIM homeobox 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: