Anti-HTR3A Rabbit Polyclonal Antibody
AVIVARP35615_P050
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine (serotonin) receptor 3A, ionotropic
- Antigen symbol:HTR3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:79246
- Antigen synonyms:5-HT3A|5-HT3|5HT3|Htr3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HTR3A (5-hydroxytryptamine (serotonin) receptor 3A, ionotropic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: