Anti-CACNB4 Rabbit Polyclonal Antibody
AVIVARP34959_P050
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, beta 4 subunit
- Antigen symbol:CACNB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:785
- Antigen synonyms:EJM6|CAB4|EA5|EIG9|CACNLB4|EJM|EJM4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CACNB4 (calcium channel, voltage-dependent, beta 4 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: