Anti-TRIM69 Rabbit Polyclonal Antibody
AVIVARP34765_P050
- Antibody type:Primary
- Antigen name:tripartite motif containing 69
- Antigen symbol:TRIM69
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:140691
- Antigen synonyms:HSD-34|Trif|HSD34|RNF36
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TRIM69 (tripartite motif containing 69)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat