Order Entry
Italy
ContactUsLinkComponent
 
Anti-A1BG Rabbit Polyclonal Antibody
 
undefined
Anti-A1BG Rabbit Polyclonal Antibody

 

  • Catalog No:
  • AVIVARP33810_P050
  • AVIVARP33810_T100
  • Antigen symbol:
  • A1BG
  • A1BG
  • Antigen name:
  • Alpha-1-B Glycoprotein
  • Alpha-1-B Glycoprotein
  • Antigen synonyms:
  • GAB|HYST2477|A1B|ABG
  • GAB|HYST2477|A1B|ABG
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 1
  • 1
  • Western blot:
  • Yes
  • Yes
  • Epitope:
  • N-terminal
  • N-terminal
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together