Order Entry
Italy
ContactUsLinkComponent
Anti-SOX12 Rabbit Polyclonal Antibody
Anti-SOX12 Rabbit Polyclonal Antibody
Certificates
undefined
Anti-SOX12 Rabbit Polyclonal Antibody

Specifications

  • Catalog No:
  • AVIVARP33330_P050
  • AVIVARP33330_T100
  • Antigen symbol:
  • SOX12
  • SOX12
  • Antigen name:
  • SRY (sex determining region Y)-box 12
  • SRY (sex determining region Y)-box 12
  • Antigen synonyms:
  • SOX22
  • SOX22
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Guinea pig, Dog, Rat, Mouse
  • Human, Guinea pig, Dog, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 6666
  • 6666
  • Western blot:
  • Yes
  • Yes
  • Epitope:
  • C-terminal
  • C-terminal
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together