Anti-AES Rabbit Polyclonal Antibody
AVIVARP32323_P050
- Antibody type:Primary
- Antigen name:amino-terminal enhancer of split
- Antigen symbol:AES
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:166
- Antigen synonyms:AES-1|AES-2|TLE5|Grg-5|ESP1|GRG5|GRG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: AES (amino-terminal enhancer of split)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: