Anti-CYP17A1 Rabbit Polyclonal Antibody
BOSSBS-6695R
: Bioss
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 17, Subfamily A, Polypeptide 1
- Antigen symbol:CYP17A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:steroid 17-alpha-monooxygenase,17?-hydroxylase/17,20 lyase/17,20 desmolase|CYP17|S17AH|P450C17|CPT7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: CYP17A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: