Anti-SLC2A10 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6325R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 2 Member A10
- Antigen symbol:Solute carrier family 2, member 10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:GLUT10|ATS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SLC2A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat